4-Iod-3-methyl-5-phenylisoxazol 95%

BLDPBD98085-1GEA 196 EUR
BLDPBD98085-1G BLDPBD98085-5G BLDPBD98085-250MG
4-Iod-3-methyl-5-phenylisoxazol 95%
4-Iodo-3-methyl-5-phenylisoxazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₈INO
Molekulargewicht: 285,0800 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03086126
CAS-Nummer: 16114-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR