4-Nitrobenzotriazol 97%

BLDPBD99381-100MGEA 21 EUR
BLDPBD99381-100MG BLDPBD99381-250MG BLDPBD99381-5G BLDPBD99381-1G
4-Nitrobenzotriazol 97%
4-Nitrobenzotriazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄N₄O₂
Molekulargewicht: 164,1216 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00160033
CAS-Nummer: 6299-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR