5-Fluor-2-(trifluormethyl)benzaldehyd ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L19901.06EA
384
EUR
L19901.06
5-Fluor-2-(trifluormethyl)benzaldehyd ≥97%
5-Fluor-2-(trifluormethyl)benzaldehyd
Formel:
C₈H₄F₄O Molekulargewicht: 192,11 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00236276 CAS-Nummer: 90381-08-1 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...