tri-Natriumcitrat 5,5-Hydrat ≥99,0% (Trockenmasse), EMPROVE® ESSENTIAL FU VIII, E331, SAFC®
1.06431.9029EA 0 EUR
1.06431.9029 1.06431.1000
tri-Natriumcitrat 5,5-Hydrat ≥99,0% (Trockenmasse), EMPROVE® ESSENTIAL FU VIII, E331, SAFC®
tri-Natriumcitrat 5,5-Hydrat
Formel: C₆H₅Na₃O₇·5.5H₂O
Molekulargewicht: 357,15 g/mol
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 6858-44-2
EINECS: 200-675-3
Bestellen
Spezifikation Testergebnisse
Assay (perchloric acid titration, calculated on dried substance) ≥ 99.0 %
Identity passes test
Appearance of solution passes test
pH-value (5 %; water) 7.5 - 9.0
Reaction passes test
Chloride (Cl) ≤ 0.0330 %
Oxalate (C₂O₄) ≤ 0.0100 %
Oxalic acid passes test
Sulfate (SO₄) ≤ 0.1000 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Fe (Iron) ≤ 0.001 %
Hg (Mercury) ≤ 0.0001 %
Pb (Lead) ≤ 0.0001 %
Readily carbonisable substance passes test
Loss on drying (150 °C) 26.0 - 30.0 %
Loss on drying (180 °C) ≤ 30.0 %