5-Fluor-2-indolcarbonsäure

Lieferant: Molekula

MOLE27753742-5GEA 0 EUR
MOLE27753742-5G
5-Fluor-2-indolcarbonsäure
5-Fluoroindole-2-carboxylic acid
Formel: C₉H₆FNO₂
Molekulargewicht: 179,15 g/mol
Schmelzpunkt: 258…259 °C
MDL Number: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR