4-(Trifluormethoxy)-o-toluidin

Lieferant: Molekula

Synonyme: 4-(Trifluormethoxy)-2-toluidin2-Amino-5-(trifluormethoxy)toluol2-Methyl-4-(trifluormethoxy)anilin

MOLE12533371-5GEA 0 EUR
MOLE12533371-5G
4-(Trifluormethoxy)-o-toluidin
4-(Trifluormethoxy)-o-toluidin
Formel: C₈H₈F₃NO
Molekulargewicht: 191,15 g/mol
MDL Number: MFCD01631541
CAS-Nummer: 86256-59-9
UN-Nummer: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR