2-Aminophenylboronsäure
Lieferant: Molekula
Synonyme:
(2-Aminophenyl)metaborsäure
, 2-Aminobenzolboronsäure
, Dihydroxy(2-aminophenyl)boran
MOLEM72836250EA
0
EUR
MOLEM72836250
2-Aminophenylboronsäure
2-Aminophenylboronsäure
Formel:
C₆H₈BNO₂ Molekulargewicht: 136,95 g/mol |
MDL Number:
MFCD01074645 CAS-Nummer: 5570-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...