Methylrot 0,1% (w/v) in Ethanol
|
Gefahr
|
Synonyme:
2-(4-(Dimethylamino)phenylazo)benzoesäure
38698.AEEA
22
EUR
38698.AE
Methylrot 0,1% (w/v) in Ethanol
Methylrot
Formel:
C₁₅H₁₅N₃O₂ Molekulargewicht: 269,3 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002425 CAS-Nummer: 493-52-7 EINECS: 207-776-1 UN-Nummer: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...