Tetranitroblau-Tetrazoliumchlorid, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY

Synonyme: TNBT

M-Clarity™ quality level = MQ200

T4000-1GEA 313 EUR
T4000-1G T4000-500MG T4000-250MG T4000-50MG T4000-100MG
Tetranitroblau-Tetrazoliumchlorid, Sigma-Aldrich®
Tetranitroblau-Tetrazoliumchlorid
Formel: C₄₀H₂₈Cl₂N₁₂O₁₀
Schmelzpunkt: 165 °C
MDL Number: MFCD00036338
CAS-Nummer: 1184-43-6
EINECS: 214-665-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR