Vaseline, white

Lieferant: Roth Carl

Synonyme: PetrolatumVaselin

ROTHE746.1EA 4.4 EUR
ROTHE746.1
Vaseline, white
Vaseline
Schmelzpunkt: 38…56 °C
Dichte: 0,865 g/cm³ (25 °C)
CAS-Nummer: 92045-74-4
EINECS: 295-456-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR