Bottles, wide neck, with screw cap, Azlon®
Lieferant: SciLabware
Azlon®
215-0432EA 33.3 EUR
215-0432
Bottles, wide neck, with screw cap, Azlon®
Flaschen Kunststoffflaschen
HDPE, translucent with PVDC wadded black urea screw caps.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Robust construction
Ideal for storage of liquids. Wide opening allows rapid filling and dispensing of powders, granules and pastes.