Creusets
Lieferant: Avignon
459-0018EA 117 EUR
459-0018 459-0014 459-0003 459-0000 459-0001 459-0017 459-0013 459-0002 459-0005 459-0015 459-0004 459-0016
Creusets
Tiegel
These crucibles are made from glazed porcelain.
  • Good chemical resistance (except hydrofluoric acid)
  • Max. temperature for use 1200 °C
  • Tall and short forms
Not recommended for oxides and melted alkaline carbonates.