Sie suchten nach: Stickstoff-Generatoren
22  results were found
SearchResultCount:"22"
Sort Results
Listenansicht Hybridansicht (NEU)
Bewerten Sie das Suchergebnis
Artikel-Nr: (PEAK3300671)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Solaris XE 35 generator has been engineered using membrane technology to meet the gas delivery requirements for labs using LC-MS or multiple lower flo...
UOM: 1 * 1 ST
Lieferant: PEAK SCIENTIFIC
Beschreibung: These precision nitrogen trace generators produce "zero nitrogen" on demand from a compressed air source.
Lieferant: PEAK SCIENTIFIC
Beschreibung: Precision Nitrogen 250 generator provides a constant and consistent source of nitrogen for detector make-up gas at standard/typical detection levels f...
Artikel-Nr: (PEAK10-1220)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The NG range is designed specifically for use as make up and carrier gas for GC applications, which require ultra high purity nitrogen for operation.
UOM: 1 * 1 ST
Artikel-Nr: (PEAK13-1270)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Membrane technology is at the heart of the Infinity 1031, generating laboratory-grade nitrogen on-site with an existing air supply. With the ability t...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK3300670)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Peak scientific’s Solaris XE nitrogen generator offers a reliable, compressor-free gas solution that can comfortably meet the demands of most mass spe...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK81-4000)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The Corona 1010 meet the specifications of the Thermo Scientific Corona Veo Charged Aerosol Detector. This system can be purchased as a single nitroge...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK10-9556)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Stickstoff-Generatoren, Genius 1052, B×T×H: 600×750×712 mm
UOM: 1 * 1 ST
Artikel-Nr: (PEAK10-2320)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The NG5000 delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC...
UOM: 1 * 1 ST
Artikel-Nr: (564-0285)
Lieferant: PEAK SCIENTIFIC
Beschreibung: For laboratories using any single quad LC-MS systems, the Genius SQ 24 nitrogen gas generator delivers analytical grade nitrogen supply dedicated for ...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK3300253)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Genius XE Nitrogen is a cutting-edge evolution combining advanced technology with refined and robust engineering. Genius XE Nitrogen provides a premiu...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK10-1295)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The NG5000(A) delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for...
UOM: 1 * 1 ST
Artikel-Nr: (564-0283)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Designed and engineered as an on-site, compressor less nitrogen generation solution tailored for the Shimadzu LCMS-8045, 8050 or 8060. The Infinity 10...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK61-0600)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The Precision Nitrogen 600 has been developed to provide a constant and consistent source of nitrogen for detector make-up gas at standard/typical det...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK3303147)
Lieferant: PEAK SCIENTIFIC
Beschreibung: Producing between 10 and 520 L/min of high purity nitrogen gas, the Infinity XE 60 series nitrogen membrane gas generators can comfortably supply mult...
UOM: 1 * 1 ST
Artikel-Nr: (PEAK10-9561)
Lieferant: PEAK SCIENTIFIC
Beschreibung: The Genius 1053 nitrogen generator provides high purity nitrogen at up to 20 litres per minute for laboratory applications. The Genius 1053 can supply...
UOM: 1 * 1 ST
Preis auf Anfrage
Lager für diesen Artikel ist begrenzt, kann aber in einem Lagerhaus in Ihrer Nähe zur Verfügung. Bitte stellen Sie sicher, dass Sie in sind angemeldet auf dieser Seite, so dass verfügbare Bestand angezeigt werden können. Wenn das call noch angezeigt wird und Sie Hilfe benötigen, rufen Sie uns an +43 1 97002 - 0.
Lager für diesen Artikel ist begrenzt, kann aber in einem Lagerhaus in Ihrer Nähe zur Verfügung. Bitte stellen Sie sicher, dass Sie in sind angemeldet auf dieser Seite, so dass verfügbare Bestand angezeigt werden können. Wenn das call noch angezeigt wird und Sie Hilfe benötigen, rufen Sie uns an +43 1 97002 - 0.
Dual Use Güter können nur im EU-Raum geliefert werden.
Dual Use Güter können nur im EU-Raum geliefert werden.
Dieses Produkt wurde von Ihrer Organisation gesperrt.
Dieses Produkt ist Ersatz für den von Ihnen gewünschten Artikel.
Abverkauf Bestand. Der Artikel wird aus dem Programm genommen/ersetzt. Alternative Artikel werden ggf. im Suchergebnis angezeigt oder wenden Sie sich an unseren Kundenservice.
1 - 16 of 22
no targeter for Bottom