Heizplatten, CP300
Lieferant: Cole Parmer
Stuart®
453-0000EA 4460 EUR
453-0000
Heizplatten, CP300
Heizgeräte Heizplatten
PTFE construction, glass ceramic plate. Separate control box (150×160×65 mm) connected via a PTFE coated lead. Ideal for digestions and metal analysis.
  • Exceptional resistance to chemical attack
  • BioCote® antimicrobial protection
The controller is also fitted with a 'hot' warning light that will flash whenever the plate temperature of the hotplate is above 50 °C and it will continue to operate when the hotplate is turned off and remains connected to the electricity supply.
Zertifizierungen: IP protection class according to DIN EN 60529: IP 43 (plate) and IP 30 (controller).