3-Brom-5-ethoxy-4-hydroxybenzaldehyd
Lieferant: Thermo Scientific
MAYBBTB10260.1EA
0
EUR
MAYBBTB10260.1
CHMPBTB10260.250
3-Brom-5-ethoxy-4-hydroxybenzaldehyd
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde
Formel:
C₉H₉BrO₃ |
MDL Number:
MFCD00173904 CAS-Nummer: 3111-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...