2-Nitrophenylhydrazin
Lieferant: Thermo Scientific
MAYBBTB09060.1EA
0
EUR
MAYBBTB09060.1
CHMPBTB09060.250
2-Nitrophenylhydrazin
2-Nitrophenylhydrazin
Formel:
C₆H₇N₃O₂ Molekulargewicht: 153,14 g/mol |
CAS-Nummer:
3034-19-3 EINECS: 221-222-6 UN-Nummer: 2926 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...