Methyl-5-chlor-2-methoxybenzoat

Lieferant: Molekula

Synonyme: 5-Chlor-2-methoxybenzoesäuremethylester

MOLE12080468-100GEA 131 EUR
MOLE12080468-100G
Methyl-5-chlor-2-methoxybenzoat
Methyl-5-chlor-2-methoxybenzoat
Formel: C₉H₉ClO₃
Molekulargewicht: 200,62 g/mol
MDL Number: MFCD00017511
CAS-Nummer: 33924-48-0
EINECS: 251-743-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR