1,4-Dibenzoylbutan
Lieferant: Molekula
Synonyme:
1,6-Diphenyl-1,6-butandion
, 1,6-Diphenylbutan-1,6-dion
MOLEM56222267EA
0
EUR
MOLEM56222267
1,4-Dibenzoylbutan
1,4-Dibenzoylbutan
Formel:
C₁₈H₁₈O₂ Molekulargewicht: 266,34 g/mol |
MDL Number:
MFCD00004761 CAS-Nummer: 3375-38-0 EINECS: 222-166-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...