6-Methoxysalicylsäure

Lieferant: Molekula

Synonyme: 2-Hydroxy-6-methoxybenzoesäure

MOLEM71905406EA 0 EUR
MOLEM71905406
6-Methoxysalicylsäure
6-Methoxysalicylsäure
Formel: C₈H₈O₄
Molekulargewicht: 168,15 g/mol
MDL Number: MFCD00674090
CAS-Nummer: 3147-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR