Magnetic stirrer, TK15
Lieferant: Kartell
KART2234EA 2880 EUR
KART2234
Magnetic stirrer, TK15
These magnetic stirrers are constructed with tested epoxy-coated metal structure.
  • Very good speed control even at low revolution
  • Chemical and corrosion resistant
  • Used with a thermostatic water tank
  • High strength permanent crusher magnet is used
These can hold 15 containers with a maximum diameter of 64 mm. The stirrer remains cool, even after continuous process, this feature makes them suitable for microbiology and biochemistry applications.
Maximum stirring capacity of the TK15 is 15 beakers of 250 ml.