4-Chlorsalicylsäure

Lieferant: Molekula
Achtung

MOLEM67650078EA 0 EUR
MOLEM67650078
4-Chlorsalicylsäure
4-Chlorsalicylsäure
Formel: C₇H₅ClO₃
Molekulargewicht: 172,57 g/mol
MDL Number: MFCD00002449
CAS-Nummer: 5106-98-9
EINECS: 225-829-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR