3-Chlor-2,4,5-trifluorbenzoesäure

Lieferant: Molekula

MOLEM75630559EA 0 EUR
MOLEM75630559
3-Chlor-2,4,5-trifluorbenzoesäure
3-Chloro-2,4,5-trifluorobenzoic acid
Formel: C₇H₂ClF₃O₂
Molekulargewicht: 210,54 g/mol
MDL Number: MFCD00153101
CAS-Nummer: 101513-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR