Micro-Ubbelohde viscometers with TC sensors
Lieferant: SI Analytics
SCOI285423790EA 1230 EUR
SCOI285423790 SCOI285423890 SCOI285423780 SCOI285423800 SCOI285423910 SCOI285423900 SCOI285423730 SCOI285423840 SCOI285423720 SCOI285423830 SCOI285423820 SCOI285423710 SCOI285423810 SCOI285423770 SCOI285423880 SCOI285423760 SCOI285423870 SCOI285423750 SCOI285423860 SCOI285423740 SCOI285423850
Micro-Ubbelohde viscometers with TC sensors
The micro Ubbelohde viscometers with TC sensors are suitable for measurements in liquids.
  • Opaque, black and/or electric conductive measuring samples
  • Suspended ball-level for determination of absolute and relative kinematic viscosity
  • Meniscus passage detected due to the different conductivity of the liquid phase and the gas phase
Suitable for the determination of the kinematic viscosity of all liquids with Newtonian flow behaviour.
Zertifizierungen: DIN 51562-2