4,5,6,7-Tetrafluorindol

Lieferant: Molekula

MOLE35498391-1GEA 0 EUR
MOLE35498391-1G
4,5,6,7-Tetrafluorindol
4,5,6,7-Tetrafluoroindole
Formel: C₈H₃F₄N
Molekulargewicht: 189,11 g/mol
MDL Number: MFCD00236717
CAS-Nummer: 16264-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR