Diethylethylmalonat

Lieferant: Molekula

MOLEM46927464EA 0 EUR
MOLEM46927464
Diethylethylmalonat
Diethylethylmalonat
Formel: C₉H₁₆O₄
Molekulargewicht: 188,22 g/mol
MDL Number: MFCD00009167
CAS-Nummer: 133-13-1
EINECS: 205-093-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR