1-Benzyl-4-piperidon

Lieferant: Molekula

MOLE45258492-100GEA 92.1 EUR
MOLE45258492-100G
1-Benzyl-4-piperidon
1-Benzyl-4-piperidon
Formel: C₁₂H₁₅NO
Molekulargewicht: 189,26 g/mol
MDL Number: MFCD00006192
CAS-Nummer: 3612-20-2
EINECS: 222-782-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR