1-Phenyloxindol

Lieferant: Molekula

MOLEM57801964EA 0 EUR
MOLEM57801964
1-Phenyloxindol
1-Phenyloxindol
Formel: C₁₄H₁₁NO
Molekulargewicht: 209,25 g/mol
MDL Number: MFCD00234850
CAS-Nummer: 3335-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR