Methyl-7-indolcarboxylat

Lieferant: Molekula

MOLE34073546-500MGEA 0 EUR
MOLE34073546-500MG
Methyl-7-indolcarboxylat
Methyl-7-indolcarboxylat
Formel: C₁₀H₉NO₂
Molekulargewicht: 175,19 g/mol
MDL Number: MFCD00211064
CAS-Nummer: 93247-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR