5-Brom-2-thiophencarbaldehyd

Lieferant: Molekula

MOLEM16348332EA 161 EUR
MOLEM16348332
5-Brom-2-thiophencarbaldehyd
5-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molekulargewicht: 191,05 g/mol
MDL Number: MFCD00005432
CAS-Nummer: 4701-17-1
EINECS: 225-176-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR