2-Chlorethylammoniumchlorid

Lieferant: Molekula

MOLE13713550-500GEA 0 EUR
MOLE13713550-500G
2-Chlorethylammoniumchlorid
2-Chlorethylammoniumchlorid
Formel: C₂H₇Cl₂N
Molekulargewicht: 115,99 g/mol
MDL Number: MFCD00012887
CAS-Nummer: 870-24-6
EINECS: 212-793-2
UN-Nummer: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR