4'-Hydroxy-3'-nitroacetophenon

Lieferant: Molekula

MOLE28698508-5GEA 0 EUR
MOLE28698508-5G
4'-Hydroxy-3'-nitroacetophenon
4'-Hydroxy-3'-nitroacetophenon
Formel: C₈H₇NO₄
Molekulargewicht: 181,15 g/mol
MDL Number: MFCD00017002
CAS-Nummer: 6322-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR