2-Benzo[b]thiophenboronsäure

Lieferant: Molekula

MOLEM47873272EA 205 EUR
MOLEM47873272
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Molekulargewicht: 178,02 g/mol
MDL Number: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR