8-Chinolincarbonsäure

Lieferant: Molekula
Achtung

MOLEM61054555EA 0 EUR
MOLEM61054555
8-Chinolincarbonsäure
8-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molekulargewicht: 173,17 g/mol
MDL Number: MFCD00047619
CAS-Nummer: 86-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR