TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure)

Lieferant: Molekula

MOLEM13809795EA 0 EUR
MOLEM13809795
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure)
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure)
Formel: C₇H₁₇NO₇S
Molekulargewicht: 259,28 g/mol
MDL Number: MFCD00038352
CAS-Nummer: 68399-81-5
EINECS: 269-993-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR