Bariumtrifluormethansulfonat
Lieferant: Molekula
MOLE33858797-5GEA
0
EUR
MOLE33858797-5G
Bariumtrifluormethansulfonat
Bariumtrifluormethansulfonat
Formel:
C₂BaF₆O₆S₂ Molekulargewicht: 435,45 g/mol |
MDL Number:
MFCD00143644 CAS-Nummer: 2794-60-7 EINECS: 220-526-6 UN-Nummer: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...