Industrial balances, compact bench scales, Catapult™
Lieferant: Ohaus
Catapult®
611-5325EA 140 EUR
611-5325 611-5327
Industrial balances, compact bench scales, Catapult™
Waagen
Simple large pan bench scale with 2 units either pounds or kilograms.
  • Portable - can be operated using 3× C batteries
  • ABS plastic housing and pan
  • Clear LCD display