Industrial scales, Valor™ 1000
Lieferant: Ohaus
Valor®
611-5355EA 0 EUR
OHAU80251173
Industrial scales, Valor™ 1000
The Valor 1000 is ideally suited for food preparation and portioning applications with standard features including a large, removable stainless steel weighing pan for easy cleaning, checkweighing mode for quick weight checking, and the freedom of portability, provided by its long-lasting internal rechargeable battery
  • 4 keys for intuitive use
  • LCD with blue backlight
  • AC adaptor (supplied) or internal rechargeable battery
  • ABS plastic housing with stainless steel pan
  • Adjustable levelling feet and front panel level bubble