Triethanolamin Hydrochlorid
Lieferant: Molekula
MOLE25559545-100GEA
68
EUR
MOLE25559545-100G
Triethanolamin Hydrochlorid
Triethanolamin Hydrochlorid
Formel:
C₆H₁₆ClNO₃ Molekulargewicht: 185,65 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012596 CAS-Nummer: 637-39-8 EINECS: 211-284-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...