1-(2-Chlorethyl)piperidiniumchlorid
Lieferant: Molekula
MOLE12162979-500GEA
0
EUR
MOLE12162979-500G
1-(2-Chlorethyl)piperidiniumchlorid
1-(2-Chlorethyl)piperidiniumchlorid
Formel:
C₇H₁₅Cl₂N Molekulargewicht: 184,11 g/mol |
MDL Number:
MFCD00012837 CAS-Nummer: 2008-75-5 EINECS: 217-920-5 UN-Nummer: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...