Cystaminiumdichlorid

Lieferant: Molekula

MOLE21267602-100GEA 117 EUR
MOLE21267602-100G
Cystaminiumdichlorid
Cystaminiumdichlorid
Formel: C₄H₁₂N₂S₂·2HCl
Molekulargewicht: 225,20 g/mol
MDL Number: MFCD00012905
CAS-Nummer: 56-17-7
EINECS: 200-260-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR