Dichlormalononitril

Lieferant: Molekula

MOLEM57066017EA 0 EUR
MOLEM57066017
Dichlormalononitril
Dichlormalononitril
Formel: C₃Cl₂N₂
MDL Number: MFCD00001841
CAS-Nummer: 13063-43-9
EINECS: 235-955-4
UN-Nummer: 3276
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR