DL-α-Phenylglycin

Lieferant: Molekula

MOLE16677282-500GEA 191 EUR
MOLE16677282-500G
DL-α-Phenylglycin
DL-α-Phenylglycin
Formel: C₈H₉NO₂
Molekulargewicht: 151,16 g/mol
MDL Number: MFCD00064402
CAS-Nummer: 2835-06-5
EINECS: 220-608-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR