o-Phenylendiacetonitril

Lieferant: Molekula

MOLE10908583-25GEA 0 EUR
MOLE10908583-25G
o-Phenylendiacetonitril
o-Phenylendiacetonitril
Formel: C₁₀H₈N₂
Molekulargewicht: 156,19 g/mol
MDL Number: MFCD00001905
CAS-Nummer: 613-73-0
EINECS: 210-351-3
UN-Nummer: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR