4-(4-Chlorphenyl)-4-piperidinol

Lieferant: Molekula

MOLE17365868-25GEA 0 EUR
MOLE17365868-25G
4-(4-Chlorphenyl)-4-piperidinol
4-(4-Chlorphenyl)-4-piperidinol
Formel: C₁₁H₁₄ClNO
Molekulargewicht: 211,69 g/mol
MDL Number: MFCD00006001
CAS-Nummer: 39512-49-7
EINECS: 254-479-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR