(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid

Lieferant: Molekula

MOLEM59191714EA 0 EUR
MOLEM59191714
(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid
D-Panthenol
Formel: C₉H₁₉NO₄
Molekulargewicht: 205,25 g/mol
MDL Number: MFCD00065006
CAS-Nummer: 81-13-0
EINECS: 201-327-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR