2,5-Dimercapto-1,3,4-thiadiazol

Lieferant: Molekula

MOLE45380441-100GEA 0 EUR
MOLE45380441-100G
2,5-Dimercapto-1,3,4-thiadiazol
2,5-Dimercapto-1,3,4-thiadiazole
Formel: C₂H₂N₂S₃
Molekulargewicht: 150,25 g/mol
MDL Number: MFCD00003103
CAS-Nummer: 1072-71-5
EINECS: 214-014-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR