(S)-(+)-4-Phenyl-2-oxazolidinon

Lieferant: Molekula

MOLE14801815-1GEA 0 EUR
MOLE14801815-1G
(S)-(+)-4-Phenyl-2-oxazolidinon
(S)-(+)-4-Phenyl-2-oxazolidinon
Formel: C₉H₉NO₂
Molekulargewicht: 163,18 g/mol
MDL Number: MFCD00043396
CAS-Nummer: 99395-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR