N-Benzylglycinethylester

Lieferant: Molekula

MOLE30356549-25GEA 0 EUR
MOLE30356549-25G
N-Benzylglycinethylester
N-Benzylglycinethylester
Formel: C₁₁H₁₅NO₂
Molekulargewicht: 193,25 g/mol
MDL Number: MFCD00009174
CAS-Nummer: 6436-90-4
EINECS: 229-218-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR