Vitamin K1 (Phytomenadion, Phyllochinon)
Lieferant: Molekula
MOLE49804572-1GEA
138
EUR
MOLE49804572-1G
Vitamin K1 (Phytomenadion, Phyllochinon)
Vitamin K1 (Phytomenadion, Phyllochinon)
Formel:
C₃₁H₄₆O₂ Molekulargewicht: 450,6958 g/mol |
MDL Number:
MFCD00214063 CAS-Nummer: 84-80-0 EINECS: 201-564-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...