Oxytetracyclin Hydrochlorid

Lieferant: Molekula

MOLE29241094-25GEA 133 EUR
MOLE29241094-25G
Oxytetracyclin Hydrochlorid
Oxytetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₉
Molekulargewicht: 496,90 g/mol
MDL Number: MFCD00135815
CAS-Nummer: 2058-46-0
EINECS: 218-161-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR